Monoclonal Anti-Fibronectin antibody produced in mouse
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing