![Monoclonal Anti-BrdU antibody produced in mouse clone BU-33, ascites fluid, Immunohistology Grade](/deepweb/assets/sigmaaldrich/product/images/233/038/40ddb177-7991-414c-957a-edbd5d9e2c63/640/40ddb177-7991-414c-957a-edbd5d9e2c63.jpg)
Monoclonal Anti-BrdU antibody produced in mouse
![Monoclonal Anti-BrdU antibody produced in mouse clone BU-33, ascites fluid, Immunohistology Grade](/deepweb/assets/sigmaaldrich/product/images/233/038/40ddb177-7991-414c-957a-edbd5d9e2c63/640/40ddb177-7991-414c-957a-edbd5d9e2c63.jpg)
Synonym(s):
Anti-Bromodeoxyuridine
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2