Molsidomine
Synonym(s):
N-(Ethoxycarbonyl)-3-(4-morpholino)sydnone imine, SIN-10
Empirical Formula (Hill Notation):
C9H14N4O4
CAS Number:
Molecular Weight:
242.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing