
MK-677

Synonym(s):
Crescendo, Ibutamoren mesylate, L-163191, MK-0677, MK-667, N-[1(R)-1,2-Dihydro-1-methanesulfonylspiro-3H-indole-3,4′-piperidin)-1′-yl]carbonyl-2-(phenylmethoxy)-ethyl-2-amino-2-methylpropanamide methanesulfonate
Empirical Formula (Hill Notation):
C27H36N4O5S · CH3SO3H
CAS Number:
Molecular Weight:
624.77
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1