Skip to Content
Merck
Miltefosine ≥98% (perchloric acid titration)

Miltefosine

Miltefosine ≥98% (perchloric acid titration)
Synonym(s):
Choline hexadecyl phosphate, HePC, Hexadecyl phosphocholine
Empirical Formula (Hill Notation):
C21H46NO4P
CAS Number:
Molecular Weight:
407.57

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing