Skip to Content
Merck
Mevinolin from Aspergillus sp. ≥98% (HPLC)

Mevinolin from Aspergillus sp.

Mevinolin from Aspergillus sp. ≥98% (HPLC)
Synonym(s):
2-Methyl-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester butanoic acid, 6-α-Methylcompactin, Lovastatin, Monacolin K
Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing