Skip to Content
Merck
(−)-Methyl (S)-3-bromo-2-methylpropionate 97%

(−)-Methyl (S)-3-bromo-2-methylpropionate

(−)-Methyl (S)-3-bromo-2-methylpropionate 97%
Synonym(s):
(−)-Methyl (S)-β-bromoisobutyrate
Linear Formula:
BrCH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
181.03

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing