Methylene Blue solution
Synonym(s):
Ehrlich’s reagent III, Loeffler’s Methylene Blue
Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Application
Microorganism Suitability
Technique
Showing 1-4 of 4