![Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/729/856/9188dcf4-74ba-4da2-bb5b-08b523aad995/640/9188dcf4-74ba-4da2-bb5b-08b523aad995.png)
Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
![Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/729/856/9188dcf4-74ba-4da2-bb5b-08b523aad995/640/9188dcf4-74ba-4da2-bb5b-08b523aad995.png)
Empirical Formula (Hill Notation):
C6H7ClN4O2
CAS Number:
Molecular Weight:
202.60
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2