Skip to Content
Merck
Methyl (2S)-glycidate optical purity ee: 99% (GLC), 98%

Methyl (2S)-glycidate

Methyl (2S)-glycidate optical purity ee: 99% (GLC), 98%
Synonym(s):
Methyl (S)-oxiranecarboxylate
Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09

Products

Product Category
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing