![Metalaxyl Metabolite CGA 62826 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/211/000/88308846-dcee-453b-8e9a-60378e3a2a66/640/88308846-dcee-453b-8e9a-60378e3a2a66.png)
Metalaxyl Metabolite CGA 62826
![Metalaxyl Metabolite CGA 62826 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/211/000/88308846-dcee-453b-8e9a-60378e3a2a66/640/88308846-dcee-453b-8e9a-60378e3a2a66.png)
Synonym(s):
N-(2,6-Dimethylphenyl)-N-(2-methoxyacetyl)-DL-alanine, N-(2-Methoxyacetyl)-N-(2,6-xylyl)-DL-alanine, rac-Metalaxyl carboxylic acid, CGA 62826, Metalaxyl acid
Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing