![Mesterolone for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/010/9ca63166-3ca9-4456-8f18-2f35c88685e9/640/9ca63166-3ca9-4456-8f18-2f35c88685e9.png)
Mesterolone for system suitability
![Mesterolone for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/010/9ca63166-3ca9-4456-8f18-2f35c88685e9/640/9ca63166-3ca9-4456-8f18-2f35c88685e9.png)
Synonym(s):
Mesterolone, 1α-Methylandrostan-17β-ol-3-one
Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing