Mepivacaine hydrochloride
Synonym(s):
Mepivacaine hydrochloride, 1-Methyl-2′,6′-pipecoloxylidine hydrochloride, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride
Empirical Formula (Hill Notation):
C15H22N2O · HCl
CAS Number:
Molecular Weight:
282.81
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing