![Menadione European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/330/990/32503e74-0cb1-49d3-b037-abc19494f771/640/32503e74-0cb1-49d3-b037-abc19494f771.png)
Menadione
![Menadione European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/330/990/32503e74-0cb1-49d3-b037-abc19494f771/640/32503e74-0cb1-49d3-b037-abc19494f771.png)
Synonym(s):
2-Methyl-1,4-naphthoquinone, Vitamin K3, Menadione
Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing