Melphalan hydrochloride
Synonym(s):
4-[Bis(2-chloroethyl)amino]-L-phenylalanine hydrochloride
Empirical Formula (Hill Notation):
C13H18Cl2N2O2 · HCl
CAS Number:
Molecular Weight:
341.66
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2