Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MCM-41 100nm nanoparticle
Linear Formula:
SiO
2
CAS Number:
7631-86-9
Products
Brand
Sigma-Aldrich (5)
Quality Segment
Functional Group
Pore Size (Å)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
926876
MCM-41 100nm nanoparticle,
pore size 30 Å
Expand
View Pricing
926884
MCM-41 100nm nanoparticle,
pore size 30 Å, amine functionalized
Expand
View Pricing
926698
MCM-41 100nm nanoparticle,
pore size 30 Å, 1% in water (dispersion)
Expand
View Pricing
926906
MCM-41 100nm nanoparticle,
pore size 30 Å, PEI functionalized
Expand
View Pricing
926914
MCM-41 100nm nanoparticle,
pore size 30 Å, carboxylic acid functionalized
Expand
View Pricing
926892
MCM-41 100nm nanoparticle,
pore size 30 Å, thiol functionalized
Expand
View Pricing