![MBHA Resin 200-400 mesh, extent of labeling: 0.5-1.0 mmol/g loading, 1 % cross-linked](/deepweb/assets/sigmaaldrich/product/structures/716/957/0c328f9a-1e6a-4a80-9aea-8d98b17abea9/640/0c328f9a-1e6a-4a80-9aea-8d98b17abea9.png)
MBHA Resin
![MBHA Resin 200-400 mesh, extent of labeling: 0.5-1.0 mmol/g loading, 1 % cross-linked](/deepweb/assets/sigmaaldrich/product/structures/716/957/0c328f9a-1e6a-4a80-9aea-8d98b17abea9/640/0c328f9a-1e6a-4a80-9aea-8d98b17abea9.png)
Synonym(s):
4-Methylbenzhydrylamine hydrochloride, polymer-bound, (α-Amino-α-p-xylyl hydrochloride)polystyrene crosslinked with divinylbenzene, MBHA resin, MBHA-polystyrene
Products
Product Category
Brand
Quality Segment
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing