Malvin(chloride)
Synonym(s):
Malvidin chloride 3,5-diglucoside, Malvidin-3,5-diglucoside, Malvoside
Empirical Formula (Hill Notation):
C29H35ClO17
Products
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing