![Magnesium Standard for AAS TraceCERT®, 1 g/L Mg in nitric acid (nominal concentration)](/deepweb/assets/sigmaaldrich/product/images/210/036/90cf3493-71b8-4377-ae3b-03382feb8d8b/640/90cf3493-71b8-4377-ae3b-03382feb8d8b.jpg)
Magnesium Standard for AAS
![Magnesium Standard for AAS TraceCERT®, 1 g/L Mg in nitric acid (nominal concentration)](/deepweb/assets/sigmaaldrich/product/images/210/036/90cf3493-71b8-4377-ae3b-03382feb8d8b/640/90cf3493-71b8-4377-ae3b-03382feb8d8b.jpg)
Synonym(s):
Magnesium nitrate hexahydrate
Linear Formula:
Mg(NO3)2 · 6 H2O
CAS Number:
Molecular Weight:
256.41
Products
Brand
Formula Weight
Product Line
Format
Element
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing