Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium fluoride
Synonym(s):
Sellaite
Linear Formula:
MgF
2
CAS Number:
7783-40-6
Molecular Weight:
62.30
EC Number:
231-995-1
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
378836
Magnesium fluoride,
random crystals, optical grade, ≥99.99% trace metals basis
Expand
View Pricing
394297
Magnesium fluoride,
technical grade
Expand
View Pricing
343188
Magnesium fluoride,
pieces, 3-6 mm, 99.9% trace metals basis (excluding Na)
Expand
View Pricing