L-Threonic acid hemicalcium salt
Synonym(s):
(2R,3S)-2,3,4-Trihydroxybutyric acid hemicalcium salt, L-Threonic acid calcium salt
Empirical Formula (Hill Notation):
C8H14CaO10
CAS Number:
Molecular Weight:
310.27
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing