Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Methionine sulfone
Synonym(s):
L-2-Amino-4-(methylsulfonyl)butanoic acid
Empirical Formula (Hill Notation):
C
5
H
11
NO
4
S
CAS Number:
7314-32-1
Molecular Weight:
181.21
EC Number:
230-774-7
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Physical Form
Quality Segment
Application
Optical Isomer
Amino Acid
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M0876
L-Methionine sulfone,
≥98% (TLC)
Expand
View Pricing