Lithium carbonate
Synonym(s):
Carbolithium, Carbonic acid lithium salt, Lithium carbonate
Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-12 of 12
Compare
Product Number
Description
Pricing