Skip to Content
Merck
Lithium bis(trifluoromethanesulfonyl)imide anhydrous, 99.99% trace metals basis

Lithium bis(trifluoromethanesulfonyl)imide

Lithium bis(trifluoromethanesulfonyl)imide anhydrous, 99.99% trace metals basis
Synonym(s):
Bis(trifluoromethane)sulfonimide lithium salt, Bis(trifluoromethylsulfonyl)amine lithium salt, Lithium bistrifluoromethanesulfonimidate
Linear Formula:
CF3SO2NLiSO2CF3
CAS Number:
Molecular Weight:
287.09

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing