Lithium β-hydroxypyruvate hydrate
Synonym(s):
β-Hydroxypyruvic acid lithium salt hydrate, 3-Hydroxy-2-oxopropanoic acid lithium salt, 3-Hydroxy-2-oxopropionic acid lithium salt
Linear Formula:
HOCH2COCOOLi · aq
CAS Number:
Molecular Weight:
109.99 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing