Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipoxin A
4
Synonym(s):
Lipoxin A
4
, (5S,6R,15S)-Trihydroxy-(7E,9E,11Z,13E)-Eicosatetraenoic acid
Empirical Formula (Hill Notation):
C
20
H
32
O
5
CAS Number:
89663-86-5
Molecular Weight:
352.47
EC Number:
200-578-6
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Research Area
Quality Segment
Special Grade
Natural (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
437720
Lipoxin A
4
,
Lipoxin A₄, CAS 89663-86-5, is a potent inhibitor of cytotoxic activity of human natural killer cells. Shown to be as potent as LTB4 in stimulating human neutrophils to generate superoxides.
Expand
View Pricing
L0521
Lipoxin A
4
,
ethanol solution
Expand
View Pricing