Lipoprotein, low density from human plasma
Synonym(s):
β-Lipoprotein, LDL, Low density lipoprotein
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Package Size
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing