Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Histidinol dihydrochloride
Synonym(s):
β-Aminoimidazole-4-propanol dihydrochloride, (
S
)-2-Amino-3-(4-imidazolyl)propanol dihydrochloride
Empirical Formula (Hill Notation):
C
6
H
11
N
3
O · 2HCl
CAS Number:
1596-64-1
Molecular Weight:
214.09
EC Number:
216-482-2
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Application
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
H6647
L-Histidinol dihydrochloride,
≥98% (TLC)
Expand
View Pricing