![Levofloxacin Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/177/773/83bb0da1-8f11-4fd8-8f74-d32b5f43f2ab/640/83bb0da1-8f11-4fd8-8f74-d32b5f43f2ab.png)
Levofloxacin Related Compound B
![Levofloxacin Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/177/773/83bb0da1-8f11-4fd8-8f74-d32b5f43f2ab/640/83bb0da1-8f11-4fd8-8f74-d32b5f43f2ab.png)
Synonym(s):
(S)-(−)-9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
Empirical Formula (Hill Notation):
C13H9F2NO4
CAS Number:
Molecular Weight:
281.21
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2