![Leukotriene B4 ~100 μg/mL in ethanol, ≥97%](/deepweb/assets/sigmaaldrich/product/structures/780/440/820199ab-a281-4ddf-b058-fe6ed5d9aec2/640/820199ab-a281-4ddf-b058-fe6ed5d9aec2.png)
Leukotriene B4
![Leukotriene B4 ~100 μg/mL in ethanol, ≥97%](/deepweb/assets/sigmaaldrich/product/structures/780/440/820199ab-a281-4ddf-b058-fe6ed5d9aec2/640/820199ab-a281-4ddf-b058-fe6ed5d9aec2.png)
Synonym(s):
[5S,12R]-Dihydroxy-[6Z,8E,10E,14Z]-eicosatetraenoic acid
Empirical Formula (Hill Notation):
C20H32O4
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing