L-(+)-Arabinose
Synonym(s):
L(+)-Arabinose
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Microorganism Suitability
Optical Isomer
Organoleptic Class
Special Grade
Technique