Skip to Content
Merck
Latanoprost Related Compound B pharmaceutical secondary standard, certified reference material

Latanoprost Related Compound B

Latanoprost Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate, (15S)-Latanoprost
Empirical Formula (Hill Notation):
C26H40O5
CAS Number:
Molecular Weight:
432.59

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade