Skip to Content
Merck
Lanthanum(III) chloride heptahydrate 99.999% trace metals basis

Lanthanum(III) chloride heptahydrate

Lanthanum(III) chloride heptahydrate 99.999% trace metals basis
Synonym(s):
Lanthanum(III) chloride heptahydrate 98%
Linear Formula:
LaCl3 · 7H2O
CAS Number:
Molecular Weight:
371.37

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade