Lanatoside C
Synonym(s):
Digilanide C, Digilanogen C, Isolanide, Lanatigen C
Empirical Formula (Hill Notation):
C49H76O20
CAS Number:
Molecular Weight:
985.12
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing