![Lamotrigine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/448/7b0f6c56-b28a-4e74-8708-7129023aa394/640/7b0f6c56-b28a-4e74-8708-7129023aa394.png)
Lamotrigine for peak identification
![Lamotrigine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/448/7b0f6c56-b28a-4e74-8708-7129023aa394/640/7b0f6c56-b28a-4e74-8708-7129023aa394.png)
Synonym(s):
Lamotrigine, 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X
Empirical Formula (Hill Notation):
C9H7Cl2N5
CAS Number:
Molecular Weight:
256.09
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing