Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Isoamyl benzoate
Synonym(s):
Isoamyl benzoate
Linear Formula:
C
6
H
5
CO
2
CH
2
CH
2
CH(CH
3
)
2
CAS Number:
94-46-2
Molecular Weight:
192.25
EC Number:
202-334-4
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (2)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Fragrance grade (1)
Halal (1)
Kosher (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
8.14500
Isoamyl benzoate,
for synthesis
Expand
View Pricing
W205818
Isoamyl benzoate,
≥98%, FCC, FG
Expand
View Pricing