Skip to Content
Merck
Irsogladine maleate ≥98% (HPLC)

Irsogladine maleate

Irsogladine maleate ≥98% (HPLC)
Synonym(s):
2,4-Diamino-6-(2,5-dichlorophenyl)-s-triazine maleate, 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine (2Z)-2-butenedioate (1:1), Gaslon N, MN 1695
Empirical Formula (Hill Notation):
C9H7Cl2N5 ·C4H4O4
CAS Number:
Molecular Weight:
372.16

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing