Iron(III) phosphate dihydrate
Synonym(s):
Ferric phosphate dihydrate
Linear Formula:
FePO4 · 2H2O
CAS Number:
Molecular Weight:
186.85
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing