
Ioxaglic acid impurity A

Synonym(s):
3-Amino-5-[(2-hydroxyethyl)carbamoyl]-2,4,6-triiodobenzoic acid, 5-Amino-N-(2-hydroxyethyl)-2,4,6-triiodoisophthalamic acid
Empirical Formula (Hill Notation):
C10H9I3N2O4
CAS Number:
Molecular Weight:
601.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing