Skip to Content
Merck
vibrant-m

Iopamidol impurity A

vibrant-m
Synonym(s):
5-Amino-N,N′-bis[2-hydroxy-1-(hydroxymethyl)ethyl]-2,4,6-triiodobenzene-1,3-dicarboxamide, N,N′-Bis(1,3-dihydroxy-2-propyl)-5-amino-2,4,6-triiodoisophthalamide, N,N′-Bis[2-hydroxy-1-(hydroxymethyl)ethyl]-5-amino-2,4,6-triiodoisophthalamide
Empirical Formula (Hill Notation):
C14H18I3N3O6
CAS Number:
Molecular Weight:
705.02

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing