![Hydroxytyrosol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/403/863/785fd004-8238-4c2a-be54-e58e6ce1bfb0/640/785fd004-8238-4c2a-be54-e58e6ce1bfb0.png)
Hydroxytyrosol
![Hydroxytyrosol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/403/863/785fd004-8238-4c2a-be54-e58e6ce1bfb0/640/785fd004-8238-4c2a-be54-e58e6ce1bfb0.png)
Synonym(s):
3-Hydroxytyrosol, 2-(3,4-Dihydroxyphenyl)ethanol, 3,4-Dihydroxyphenethyl alcohol, DOPET, Homoprotocatechuyl alcohol
Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing