Skip to Content
Merck
Hydrogen fluoride pyridine pyridine ~30 %, hydrogen fluoride ~70 %

Hydrogen fluoride pyridine

Hydrogen fluoride pyridine pyridine ~30 %, hydrogen fluoride ~70 %
Synonym(s):
HF-Pyridine, Olah′s reagent, PPHF, Poly(pyridine fluoride), Pyridine hydrofluoride, Pyridinium poly(hydrogen fluoride), Pyridinium polybifluoride
CAS Number:

Products

Brand
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing