Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydrofluoric acid
Products
Product Category
acids (5)
Brand
Supelco (5)
Physical Form
Product Line
Agency
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.00338
Hydrofluoric acid,
40%, for analysis EMSURE
®
ISO,Reag. Ph Eur
Expand
View Pricing
1.00334
Hydrofluoric acid,
48%, for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
1.00335
Hydrofluoric acid,
40%, Suprapur
®
Expand
View Pricing
1.01513
Hydrofluoric acid,
48%, Ultrapur
Expand
View Pricing
1.00337
Hydrofluoric acid,
38-40%, EMPLURA
®
Expand
View Pricing