![Hycon® Agar Strips SDX Sabouraud Dextrose Agar, Chlortetracycline, Chloramphenicol, for yeasts, for molds, aseptically filled, single packed of 50 units, suitable for air monitoring](/deepweb/assets/sigmaaldrich/product/images/108/074/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d/640/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d.jpg)
Hycon® Agar Strips SDX
![Hycon® Agar Strips SDX Sabouraud Dextrose Agar, Chlortetracycline, Chloramphenicol, for yeasts, for molds, aseptically filled, single packed of 50 units, suitable for air monitoring](/deepweb/assets/sigmaaldrich/product/images/108/074/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d/640/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d.jpg)
Synonym(s):
Hycon Ready-to-use Agar Strips
Products
Product Category
Brand
Physical Form
Product Line
Chemical Composition
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterilization Method
Showing 1-1 of 1