Skip to Content
Products
Cart
714
AT
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Account
Order Lookup
Quick Order
Cart
714
Back
HPTLC plates, Silica gel 60 RP-18 F₂₅₄s
Products
Product Category
plates (2)
Brand
Supelco (2)
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.13724
HPTLC plates, Silica gel 60 RP-18 F₂₅₄s,
pkg of 25 plates, plate L × W 10 cm × 10 cm, glass support
Expand
View Pricing
1.16225
HPTLC plates, Silica gel 60 RP-18 F₂₅₄s,
pkg of 25 plates, plate L × W 20 cm × 10 cm, glass support
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
Deutsch - DE
English - EN
Learn More