Hexanoic acid
Synonym(s):
Acid C6, Caproic acid, Hexanoic acid
Linear Formula:
CH3(CH2)4COOH
CAS Number:
Molecular Weight:
116.16
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing