
Hexafluorozirconic acid solution

Synonym(s):
Dihydrogen hexafluorozirconate solution, Hydrogen zirconium fluoride
Linear Formula:
H2ZrF6
CAS Number:
Molecular Weight:
207.23
Products
Brand
Formula Weight
Quality Segment
Reagent Type
Core
Showing 1-1 of 1