Guanosine
Synonym(s):
9-(β-D-Ribofuranosyl)guanine, Guanine-9-β-D-ribofuranoside, Guanosine
Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4