![Guanidine Hydrochloride, Molecular Biology Grade Powerful protein denaturant.](/deepweb/assets/sigmaaldrich/product/images/363/023/09bc3ef9-4359-4468-939e-30d273ba088f/640/09bc3ef9-4359-4468-939e-30d273ba088f.jpg)
Guanidine Hydrochloride, Molecular Biology Grade
![Guanidine Hydrochloride, Molecular Biology Grade Powerful protein denaturant.](/deepweb/assets/sigmaaldrich/product/images/363/023/09bc3ef9-4359-4468-939e-30d273ba088f/640/09bc3ef9-4359-4468-939e-30d273ba088f.jpg)
Synonym(s):
Guanidinium chloride
Empirical Formula (Hill Notation):
CH5N3 · xHCl
CAS Number:
Molecular Weight:
59.07 (free base basis)
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing