Skip to Content
Merck
Glyburide (Glibenclamide) Pharmaceutical Secondary Standard; Certified Reference Material

Glyburide (Glibenclamide)

Glyburide (Glibenclamide) Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Glybenclamide, 5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glyburide, N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N′-cyclohexylurea
Empirical Formula (Hill Notation):
C23H28ClN3O5S
CAS Number:
Molecular Weight:
494.00
EC Number:

Products

Brand
Formula Weight
Gene Alias
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1